Learn More
Minigrip™ Resealable Write-on Bags
Product Code. 10634185
Click to view available options
Width (Metric):
40 mm
75 mm
100 mm
140 mm
190 mm
Height (Metric):
65 mm
85 mm
140 mm
190 mm
Unit Size:
Pack of 100
Pack of 1000
Product Code. 10634185
Brand: Minigrip™ BAJ340003N
Edge
Label bag contents with ease. Minigrip™ Resealable Write-on Bags provide three white bars specifically designed for writing on with inkpen or marker.
  • Polyethylene
  • Resealable
  • Autoclavable
  • Three write-on bars for labelling
Specifications
Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pack
Width (Metric) 40 mm
Height (Metric) 65 mm