Learn More
Christeyns™ Chriox Spray
Click to view available options
Size:
750 mL
5 L
Unit Size:
4.5 L
5 L
Description
- Eco-detergent certified by ECOCERT Greenlife according to the ECOCERT standard
- Suitable for cleaning all washable surfaces (including in the food industry)
- Spray the pure product on the surface to be treated or preferably on a clean, nonwoven cloth, and leave on for 5 to 15 min.
Specifications
Specifications
| Product Type | Surface Disinfectant |
| Color-coded | No |
| Form | Clear Liquid |
| pH | Neutral |
| Scent | Unscented |
| Sterility | Non-sterile |
| Quantity | 2 Cans |
| Size | 5 L |