Learn More
Tocris Bioscience™ CHPG Sodium salt
GREENER_CHOICE
Product Code. 18005272
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18005272
Brand: Tocris Bioscience™ 3695/10
Selective mGlu5 agonist. Sodium salt of CHPG (Cat. No. 1049)
CHPG Sodium salt is a sodium salt of the selective mGlu5 agonist CHPG (Cat. No. 1049).
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Product Type CHPG Sodium salt
Purity >99%
Molecular Weight (g/mol) 223.59
Inhibitors CHPG Sodium salt