Learn More
Chondroitin Sulfate C Sodium Salt (Technical Grade), TRC
Product Code. 17918004
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17918004 5 g 5g
17959914 25 g 25g
17977254 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17918004 Supplier Toronto Research Chemicals Supplier No. TRCC4333785G
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 12678-07-8
Molecular Formula (C14 H19 N O14 S . 2 Na)n
Molecular Weight (g/mol) 1390.1
Synonym Chondoitrin 6-sulfate sodium salt, Sodium chondroitin sulfate C
IUPAC Name 3-[3-acetamido-4-[5-[3-acetamido-4-[5-[3-acetamido-4,5-dihydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-4,5,6-trihydroxyoxane-2-carboxylate
SMILES [Na+].[Na+].[*]O[C@@H]1C(O)C(O)[C@H](O[C@@H]2C(O)C(NC(=O)C)[C@H]([*])OC2COS(=O)(=O)[O-])OC1C(=O)[O-]
Specifications
Quantity 5 g
InChI Formula InChI=1S/C42H65N3O43S3/c1-7(46)43-13-19(52)16(49)10(4-75-89(66,67)68)78-38(13)84-29-21(54)24(57)41(87-32(29)35(61)62)83-27-15(45-9(3)48)40(80-12(18(27)51)6-77-91(72,73)74)86-30-22(55)25(58)42(88-33(30)36(63)64)82-26-14(44-8(2)47)39(79-11(17(26)50)5-76-90(69,70)71)85-28-20(53)23(56)37(65)81-31(28)34(59)60/h10-33,37-42,49-58,65H,4-6H2,1-3H3,(H,43,46)(H,44,47)(H,45,48)(H,59,60)(H,61,62)(H,63,64)(H,66,67,68)(H,69,70,71)(H,72,73,74)/p-6
Formula Weight 499.0608
Chemical Name or Material Chondroitin sulfate C sodium salt