Learn More
Choline-d9 Chloride (N,N,N-trimethyl-d9), CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | Choline-D9 Chloride |
| CAS | 61037-86-3 |
| Molecular Formula | C5 H5 D9 N O . Cl |
| Quantity | 1 g |
| Synonym | 2-Hydroxy-N,N,N-trimethylethanaminium-d9 Chloride, (Trimethyl-d9)(2-hydroxyethyl)ammonium Chloride, Actiron CC 6-d9, Bilineurin-d9 Chloride, Biocolina-d9, Hepacholine-d9, Lipotril-d9, Neocolina-d9, Paresan-d9; |
| InChI Formula | InChI=1 S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1 H/q+1;/p-1/i1D3,2D3,3D3; |
| SMILES | C([N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])CO.[Cl-] |
| IUPAC Name | 2-hydroxy-N,N,N-tris(methyl-d3)ethan-1-aminium chloride |
| Molecular Weight (g/mol) | 148.68 |
| Formula Weight | 148.13 g/mol |
| Show More |