Learn More
Choline chloride, 98+%
Product Code. p-3345782
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422508 100 g 100g
11432508 500 g 500g
11442508 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11422508 Supplier Thermo Scientific Alfa Aesar Supplier No. A15828.22
Add to basket
CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
Solubility
Soluble in water, ethanol, methanol and alcohol.
Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCO
Specifications
Melting Point ∽305°C (decomposition)
Assay Percent Range ≥98%
Quantity 100 g
Beilstein 3563126
Sensitivity Hygroscopic
Merck Index 14,2206
Solubility Information Soluble in water,ethanol,methanol and alcohol.
Formula Weight 139.63
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Choline Chloride