Learn More
Choline chloride, 98+% (dry basis)
Product Code. 15825108 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 lb.
10 kg
Packaging:
Plastic bottle
Unit Size:
10kg
2268g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15825108 5 lb. Plastic bottle 2268g
15815268 10 kg Plastic bottle 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15825108 Supplier Thermo Scientific Supplier No. J13410.P5
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
SMILES [Cl-].C[N+](C)(C)CCO
Specifications
Melting Point ∼305°C (decomposition)
Packaging Plastic bottle
Quantity 5 lb.
Beilstein 3563126
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
Formula Weight 139.62
Percent Purity ≥98%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material Choline chloride