Learn More
Choline-1,1,2,2-d4 Bromide, TRC
Product Code. 30360370
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360370 10 mg -
30356115 100 mg 100mg
30358919 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30360370 Supplier Toronto Research Chemicals Supplier No. TRCC21105110MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 285979-69-3
Molecular Formula C5 H10 D4 N O . Br
Molecular Weight (g/mol) 188.09941
Synonym Ethan-1,1,2,2-d4-aminium, 2-hydroxy-N,N,N-trimethyl-, bromide (9CI)
IUPAC Name trimethyl-(1,1,2,2-tetradeuterio-2-hydroxy-ethyl)ammonium;bromide
SMILES [Br-].OCC[N+](C)(C)C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C5H14NO.BrH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i4D2,5D2;
Formula Weight 187.051
Chemical Name or Material Choline-1,1,2,2-D4 Bromide