Learn More
Choline-1,1,2,2-d4 Bromide, CDN
Product Code. 30323224
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323224 0.5 g 0.5g
30321322 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323224 Supplier CDN Supplier No. CDND16000.5G
 Restricted Item
Estimated Shipment: 10-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Choline-1,1,2,2-D4 Bromide
CAS 285979-69-3
Molecular Formula C5 H10 D4 N O . Br
Quantity 0.5 g
Synonym Ethan-1,1,2,2-d4-aminium, 2-hydroxy-N,N,N-trimethyl-, bromide (9 CI)
InChI Formula InChI=1 S/C5H14NO.BrH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1 H/q+1;/p-1/i4D2,5D2;
SMILES [Br-].OCC[N+](C)(C)C
IUPAC Name trimethyl-(1,1,2,2-tetradeuterio-2-hydroxy-ethyl)ammonium;bromide
Molecular Weight (g/mol) 188.0994
Formula Weight 187.051 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity