Learn More
Choline-1,1,2,2-d4 Bromide, CDN
Product Code. 30321322
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321322
Brand: CDN CDND16001G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Choline-1,1,2,2-D4 Bromide
CAS 285979-69-3
Molecular Formula C5 H10 D4 N O . Br
Quantity 1 g
Synonym Ethan-1,1,2,2-d4-aminium, 2-hydroxy-N,N,N-trimethyl-, bromide (9 CI)
InChI Formula InChI=1 S/C5H14NO.BrH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1 H/q+1;/p-1/i4D2,5D2;
SMILES [Br-].OCC[N+](C)(C)C
IUPAC Name trimethyl-(1,1,2,2-tetradeuterio-2-hydroxy-ethyl)ammonium;bromide
Molecular Weight (g/mol) 188.0994
Formula Weight 187.051 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity