Learn More
Cholesterol 3-Acetate, TRC
Product Code. 30359089
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359089 1 g 1g
30356141 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30359089 Supplier Toronto Research Chemicals Supplier No. TRCC4325051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 604-35-3
Molecular Formula C29 H48 O2
Molecular Weight (g/mol) 428.69
Synonym Cholest-5-en-3-ol (3β)-, 3-acetate, Cholest-5-en-3-ol (3β)-, acetate (9CI), Cholesterol, acetate (6CI,7CI,8CI), (-)-Cholesteryl acetate, (3β)-Cholest-5-en-3-ol acetate, 3-Cholesteryl acetate, 3β-Acetoxycholest-5-ene, Cholest-5-en-3β-ol acetate, Cholest-5-en-3β-yl acetate, Cholesterin acetate, Cholesterol 3-acetate, Cholesterol 3β-acetate, Cholesteryl acetate, NSC 8799
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C29H48O2/c1-19(2)8-7-9-20(3)25-12-13-26-24-11-10-22-18-23(31-21(4)30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-20,23-27H,7-9,11-18H2,1-6H3/t20-,23+,24+,25-,26+,27+,28+,29-/m1/s1
Formula Weight 428.3654
Chemical Name or Material Cholesteryl Acetate