missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholesterol-26,26,26,27,27,27-d6, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Cholesterol-26,26,26,27,27,27-D6 |
| CAS | 60816-17-3 |
| Molecular Formula | C27 H40 D6 O |
| Quantity | 0.05 g |
| Synonym | Cholest-5-en-26,26,26,27,27,27-d6-3-ol, (3β)- (9 CI, ACI), (3β)-Cholest-5-en-26,26,26,27,27,27-d6-3-ol (ACI), 26,26,26,27,27,27-Hexadeuterocholesterol, [26,26,26,27,27,27-2H6]-Cholesterol, Cholesterol-26,26,26,27,27,27-d6, Cholesterol-d6, Cholesterol-26,26,26,27,27,27-D6 |
| InChI Formula | InChI=1 S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28 H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3 |
| SMILES | C[C@@]12 [C@](CC[C@]2([H])[C@H](C)CCCC(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])([H])[C@@]3([H])[C@@](CC1)([H])[C@@]4(C(C[C@H](CC4)O)=CC3)C |
| IUPAC Name | (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-((R)-6-(methyl-d3)heptan-2-yl-7,7,7-d3)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 392.6905 |
| Formula Weight | 392.3925 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction