missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX CHO-K1 EDG4 Gq Cell Line

Product Code. 17374739
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17374739 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17374739 Supplier DiscoverX Supplier No. 951012C2
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The CHO-K1 EDG4 Gq Cell Line measures EDG4 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.

The EDG4 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. LPA) based activation of EDG4 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses EDG4 , a naturally Gq coupled GPCR. Upon activation of EDG4 , the resulting calcium activity can be fluorescently assayed using the Calcium NoWash PLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Calcium Signaling
Cell Line CHO-K1
For Use With (Application) Cell Lines
Primary Agonist 1-Oleoyl-LPA
Quantity 2 Vials
Target Entry EDG4
Accession Number NP_004711
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.