missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX CHO-K1 EDG3 Gq Cell Line
Description
The EDG3 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. S1P) based activation of EDG3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses EDG3 , a naturally Gq coupled GPCR. Upon activation of EDG3 , the resulting calcium activity can be fluorescently assayed using the Calcium NoWash PLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Calcium Signaling |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Sphingosine 1-phosphate |
| Quantity | 2 Vials |
| Target Entry | EDG3 |
| Accession Number | NM_005226 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?