missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX CHO-K1 ADRA1A Gq Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The ADRA1A Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRA1A , a naturally Gq coupled GPCR. Upon activation of ADRA1A , the resulting calcium activity can be fluorescently assayed using the Calcium NoWash PLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Calcium Signaling |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | A 61603 |
| Quantity | 2 Vials |
| Target Entry | ADRA1A |
| Accession Number | NM_000680 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction