missing translation for 'onlineSavingsMsg'
Learn More

Chlorpheniramine-d4 Maleate Salt, TRC

Product Code. 30360900
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360900 1 mg 1mg
30351562 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30360900 Supplier Toronto Research Chemicals Supplier No. TRCC4243041MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20H19D4ClN2O4
Molecular Weight (g/mol) 394.89
Synonym 2-[p-Chloro-α-(2-dimethyl-aminoethyl-d4)benzyl]pyridine Maleate Salt, 1-(p-Chlorophenyl)-1-(2-pyridil)-3-dimethylaminopropane-d4 Maleate Salt, Allergisan-d4, Piriton-d4, Teldrin-d4
IUPAC Name (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-1,1,2,2-tetradeuterio-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine
SMILES ClC(C=C1)=CC=C1C(C([2H])([2H])C([2H])([2H])N(C)C)C2=CC=CC=N2.O=C(O)/C=C\C(O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i10D2,12D2;
Chemical Name or Material Chlorpheniramine-d4 Maleate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.