Learn More
Chloroiridic Acid, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Chloroiridic Acid |
| CAS | 110802-84-1 |
| Molecular Formula | H2IrCl6.x(H2O) |
| Quantity | 250 mg |
| Synonym | Dihydrogen Hexachloroiridate(IV) Hydrate, Hydrogen Hexachloroiridate(IV) Hydrate |
| InChI Formula | InChI=1S/6ClH.Ir.H/h6*1H;;/q;;;;;;+5;/p-5 |
| SMILES | Cl[IrH](Cl)(Cl)(Cl)(Cl)[ClH] |
| IUPAC Name | pentachloro-hydrido-λ^{2}-chloranyliridium |
| Formula Weight | 404.7917 |