missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroiridic Acid, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Chloroiridic Acid |
| CAS | 110802-84-1 |
| Molecular Formula | H2IrCl6.x(H2O) |
| Quantity | 500 mg |
| Synonym | Dihydrogen Hexachloroiridate(IV) Hydrate, Hydrogen Hexachloroiridate(IV) Hydrate |
| InChI Formula | InChI=1S/6ClH.Ir.H/h6*1H;;/q;;;;;;+5;/p-5 |
| SMILES | Cl[IrH](Cl)(Cl)(Cl)(Cl)[ClH] |
| IUPAC Name | pentachloro-hydrido-λ^{2}-chloranyliridium |
| Formula Weight | 404.7917 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction