Learn More
Chloroform, BAKER ANALYZED™, J.T. Baker™
Product Code. 10086100 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10086100
Brand: JT Baker 7386.2500
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Laboratory Reagent
Chemical Identifiers
CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym Trichloromethane
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl
Specifications
Melting Point -63°C
Color Max. 10 (APHA)
Density 1.4832 g/cm³ (20°C)
Boiling Point 61.7°C (1013 hPa)
Quantity 2.5 L
Packaging Glass Bottle
Assay Percent Range min. 99%
Residue after Evaporation max. 0.001%
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P281-Use personal protective equipment as required.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.