Learn More
Chlorodimethyl-n-octylsilane, 97%
Product Code. 11369235
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369235 25 g 25g
11359235 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11369235 Supplier Thermo Scientific Alfa Aesar Supplier No. L16564.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 18162-84-0 | C10H23ClSi | 206.829 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18162-84-0
Molecular Formula C10H23ClSi
Molecular Weight (g/mol) 206.829
MDL Number MFCD00000511
InChI Key DBKNGKYVNBJWHL-UHFFFAOYSA-N
Synonym n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc
PubChem CID 87482
IUPAC Name chloro-dimethyl-octylsilane
SMILES CCCCCCCC[Si](C)(C)Cl
Specifications
Boiling Point 222°C to 225°C
Quantity 25 g
UN Number UN2987
Beilstein 1738376
Flash Point 92°C (198°F)
Formula Weight 206.83
Percent Purity 97%
Odor Acrid
Refractive Index 1.435
Sensitivity Moisture sensitive
Density 0.873
Chemical Name or Material Chlorodimethyl-n-octylsilane