Learn More
Chloroacetic Acid-d3, CDN
Specifications
Specifications
| Chemical Name or Material | Chloroacetic Acid-d3 |
| CAS | 1796-85-6 |
| Molecular Formula | C2 D3 Cl O2 |
| Quantity | 0.5 g |
| Synonym | Acetic-d2 acid-d, chloro- (7 CI, 8 CI, 9 CI), Trideuteriochloroacetic acid, Chloroacetic-d2 acid-d, Chloroacetic acid-D3, Chloroacetic Acid-d3 |
| InChI Formula | InChI=1 S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)/i1D2/hD |
| SMILES | [2 H]OC(=O)C([2 H])([2 H])Cl |
| IUPAC Name | deuterio 2-chloro-2,2-dideuterioacetate |
| Molecular Weight (g/mol) | 97.5155 |
| Formula Weight | 97.001 g/mol |
| Show More |