Learn More
Chlormezanone, TRC
Product Code. 30361419
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361419 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30361419 Supplier Toronto Research Chemicals Supplier No. TRCC60201525MG
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlormezanone
CAS 80-77-3
Molecular Formula C11 H12 Cl N O3 S
Quantity 25 mg
Synonym Chlormezanone, 4H-1,3-Thiazin-4-one, 2-(4-chlorophenyl)tetrahydro-3-methyl-, 1,1-dioxide, 4H-1,3-Thiazin-4-one, 2-(p-chlorophenyl)tetrahydro-3-methyl-, 1,1-dioxide (6CI,7CI,8CI), (±)-Chlormezanone, (±)-Fenarol, 2-(4-Chlorophenyl)-3-methyl-4-metathiazanone-1,1-dioxide, 2-(p-Chlorophenyl)-3-methyl-1,3-perhydrothiazin-4-one 1,1-dioxide, 2-(p-Chlorophenyl)tetrahydro-3-methyl-4H-1,3-thiazin-4-one 1,1-dioxide, 2-(p-Chlorphenyl)-3-methyl-1,3-perhydrothiazin-4-on-1,1-dioxide, Alinam, Banabin-Sintyal, Chlormethazanone, Chlormethazone, Dichloromezanone, Fenarol, Lobak, Mio-Sed, Muskel Trancopal, NSC 169108, Phenarol, Rexan, Rilansyl, Rilaquil, Rilasol, Rilassol, Supotran, Suprotran, Tanafol, Trancopal, Trancote, Transanate, dl-Chlormezanone
InChI Formula InChI=1S/C11H12ClNO3S/c1-13-10(14)6-7-17(15,16)11(13)8-2-4-9(12)5-3-8/h2-5,11H,6-7H2,1H3
SMILES CN1C(c2ccc(Cl)cc2)S(=O)(=O)CCC1=O
IUPAC Name 2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
Molecular Weight (g/mol) 273.74
Formula Weight 273.0226