Learn More
Lovibond™ Chlorine reference standard kit for MD100
Designed to check the accuracy and the reliability of the results
Brand: Lovibond™ 275650
Additional Details : Weight : 0.32000kg
Description
Designed to check the accuracy and the reliability of the results
It is not possible to calibrate the photometer with the reference standards.The shelf life of reference standards is two years from the date of production, provided that storage and use are in accordance with the instructions provided.
Specifications
measures 0.2 mg/L and 1.0 mg/L standards chlorine | |
Chlorine Standard |