Learn More
Lovibond™ Chlorine reference standard kit for MD100
Designed to check the accuracy and the reliability of the results
84.10€ - 106.00€
Specifications
Type Chlorine Standard
View More Specs
Products 2
Product Code Brand Range Price Quantity & Availability  
Product Code Brand Range Price Quantity & Availability  
15795912
View Documents Promotion Details
Lovibond™
275650
measures 0.2 mg/L and 1.0 mg/L standards chlorine
106.00€
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
15610017
View Documents Promotion Details
Lovibond™
275656
measures 1.0 mg/L and 4.0 mg/L standards chlorine
84.10€
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
Description
Designed to check the accuracy and the reliability of the results
It is not possible to calibrate the photometer with the reference standards.
The shelf life of reference standards is two years from the date of production, provided that storage and use are in accordance with the instructions provided.
Specifications
Specifications
Chlorine Standard