Learn More
Lovibond™ Chlorine reference standard kit for MD100
Designed to check the accuracy and the reliability of the results
84.10€ - 106.00€
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
15795912
|
Lovibond™
275650 |
measures 0.2 mg/L and 1.0 mg/L standards chlorine |
106.00€
1 set |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
15610017
|
Lovibond™
275656 |
measures 1.0 mg/L and 4.0 mg/L standards chlorine |
84.10€
1 set |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
Designed to check the accuracy and the reliability of the results
It is not possible to calibrate the photometer with the reference standards.The shelf life of reference standards is two years from the date of production, provided that storage and use are in accordance with the instructions provided.
Specifications
Chlorine Standard |