Learn More
Hanna Instruments™ Chlorine Liquid Reagents
Código de producto. 11769392
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Type:
Chlorine Reagent
Liquid Reagent Kit
missing translation for 'unitSize'
Pack of 300
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Type unitSize
11769392 Chlorine Reagent Pack of 300
11759392 Liquid Reagent Kit Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11769392 missing translation for 'mfr' Hanna Instruments™ missing translation for 'supplierNo' HI93701T
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Ver productos alternativos
Este artículo no se puede devolver. Vea la política de devoluciones
Reagents for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers.
  • Reagents for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers
  • High quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements
  • Reagents follow an adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G
  • The methods use a DPD reagent in which the reaction between chlorine and reagent causes a pink tint in the sample
  • Pre-made reagents for ease of use
  • Prepared with high purity chemicals
Especificaciones
No. of Tests 300
Quantity 1 Each
Range 0 to 5 mg/L
Test Method adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G
Type Chlorine Reagent
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • X300 LIQUID TOT CHLORINE REAGENT
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Indicaciones de peligro
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
  • P390-Absorb spillage to prevent material damage.
Otros peligros
  • MIXTURE LIST-Contain: sulphuric acid, sodium hydroxide, potassium iodide
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.