Learn More
Hanna Instruments™ Chlorine Liquid Reagents
Descripción
- Reagents for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers
- High quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements
- Reagents follow an adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G
- The methods use a DPD reagent in which the reaction between chlorine and reagent causes a pink tint in the sample
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
Especificaciones
Especificaciones
| No. of Tests | 300 |
| Quantity | 1 Each |
| Range | 0 to 5 mg/L |
| Test Method | adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G |
| Type | Chlorine Reagent |
Seguridad y manipulación
Nombre del producto
- X300 LIQUID TOT CHLORINE REAGENT
- Danger
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362-Take off contaminated clothing.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contain: sulphuric acid, sodium hydroxide, potassium iodide
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?