Learn More
Hanna Instruments™ Chlorine Liquid Reagents
Product Code. 11759392
Change view
Click to view available options
Type:
Chlorine Reagent
Liquid Reagent Kit
Unit Size:
Pack of 300
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11759392 Liquid Reagent Kit Pack of 300
11769392 Chlorine Reagent Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11759392 Supplier Hanna Instruments™ Supplier No. HI93701F
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Reagents for the colorimetric determination of total and free chlorine, for use with Hanna’s compatible benchtop and portable photometers.
  • Reagents for the colorimetric determination of free chlorine, for use with Hanna’s line of compatible benchtop and portable meters 
  • Pre-made reagents for ease of use
  • Prepared with high purity chemicals
  • High quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements
  • Reagents follow the DPD Method in which the reaction between chlorine and reagent causes a pink tint in the sample
Specifications
Analytes Chlorine (Free) Reagent
No. of Tests 300
Packaging Dropper Bottle
Range 0 to 5 mg/L
Test Method Adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G
Type Liquid Reagent Kit
compliance-icons
Product Identifier
  • X300 LIQUID FREE CHLORINE REAGENT
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid, sodium hydroxide, N,N-diethyl-1,4-phenylenediammonium sulphate