Learn More
Hanna Instruments™ Chlorine Liquid Reagents
Description
- Reagents for the colorimetric determination of free chlorine, for use with Hanna’s line of compatible benchtop and portable meters
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
- High quality liquid reagents that allow for users to achieve fast and accurate colorimetric measurements
- Reagents follow the DPD Method in which the reaction between chlorine and reagent causes a pink tint in the sample
Specifications
Specifications
| Analytes | Chlorine (Free) Reagent |
| No. of Tests | 300 |
| Packaging | Dropper Bottle |
| Range | 0 to 5 mg/L |
| Test Method | Adaptation of the USEPA method 330.5 and Standard Method 4500-Cl G |
| Type | Liquid Reagent Kit |
Safety and Handling
Product Identifier
- X300 LIQUID FREE CHLORINE REAGENT
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contain: sulphuric acid, sodium hydroxide, N,N-diethyl-1,4-phenylenediammonium sulphate