missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Chlorine Dioxide Reagent Sets, For Colorimetric Water Testing

Product Code. 11873663
Change view
Click to view available options
Range:
0.00 to 2.00 mg/L Chlorine Dioxide
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
11873663 0.00 to 2.00 mg/L Chlorine Dioxide Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11873663 Supplier Hanna Instruments™ Supplier No. HI9373801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Dioxide Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.

These high quality reagents are designed to follow an adaptation of the Chlorophenol Red Method for colorimetric determination of Chlorine Dioxide. By simply adding the HI93738-01 reagents to the sample, a reaction will take place and a compatible photometer can be used to determine the concentration from the purple color that is produced.

  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability

Specifications

Analytes Chlorophenol Red
For Use With (Application) Chlorine dioxide determination
For Use With (Equipment) Compatible Hanna benchtop and portable photometers (HI83200, HI83205, HI83099, HI96738)
Includes Dropper bottles and individual packets
No. of Tests 100
Quantity 1 Set
Range 0.00 to 2.00 mg/L Chlorine Dioxide
Test Method Cholorimetric determination
Type Water Testing Kit
compliance-icons
Product Identifier
  • Reagents for 100 tests chlorine dioxide .
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Aminomethyl Propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.