missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CHIR98014

Product Code. 17160162
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17160162

Brand: STEMCELL Technologies 73044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

WNT pathway activator; Inhibits GSK3α and GSK3β

  • CHIR98014 is a reversible, cell-permeable activator of the WNT pathway, through inhibition of both isoforms of glycogen synthase kinase 3 (GSK3α and GSK3β) with IC50 values of 0.65 and 0.58 nM, respectively
  • It shows at least 500-fold selectivity for GSK-3 versus 20 other serine/threonine or tyrosine kinases (Ring et al.)

Specifications

CAS 556813-39-9
Quantity 10 mg
Molecular Formula C20H17Cl2N9O2
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.