missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chenodeoxycholic acid, 98%

Product Code. 10368613 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10368613

Brand: Thermo Scientific Chemicals 228771000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chenodeoxycholic acid, 98%, ACROS Organics, Quantity: 5g, Packaging: Glass bottle, White, Melting Point: 162 deg.C to 168 deg.C, Molecular Weight: 391.572, Percent Purity: >-97.5%, Assay Percent Range: 97+%, CAS: 474-25-9, ChEBI: CHEBI:36234, EINECS Number: 207-481-8, Formula Weight: 392.57

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474-25-9
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00064142
InChI Key RUDATBOHQWOJDD-BSWAIDMHSA-N
Synonym chenodexycholic acid, chenodeoxycholate anion, chenodeoxycholate 1-, 3alpha, 7alpha-dihydroxy-5beta-cholanic acid, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oate, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oic acid anion, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-10,13-dimethyl-3,7-bis oxidanyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate
PubChem CID 9548590
ChEBI CHEBI:36234
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Melting Point 162°C to 168°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 100 g
Specific Rotation Condition +11.50 (20.00°C c=1,EtOH)
Specific Rotation +10° to +13° (20°C, 589nm) (c=1, EtOH)
Formula Weight 392.57
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Chenodeoxycholic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.