Learn More
Saint Gobain Life Sciences Chemware™ PFA Graduated Erlenmeyer Flask
Product Code. p-8369102
Click to view available options
Capacity (Metric):
100 mL
200 mL
300 mL
Unit Size:
Each
Product Code. 11724628
Brand: Saint Gobain Life Sciences D1069075
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemware™ PFA Graduated Erlenmeyer Flask is chemically inert for ultra-pure applications.
  • Graduated
  • Transparent for easy viewing
  • Non-stick PFA surface-easy to clean
  • Can be used in a microwave
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 3.4 oz.
Color Transparent
Diameter (Metric) Outer Neck 23 mm
Capacity (Metric) 100 mL
Material PFA
Height (English) 3.8 in.
Height (Metric) 97 mm
Product Type Erlenmeyer Flask
Graduated Yes