Learn More
Saint Gobain Life Sciences Chemware™ PFA Graduated Erlenmeyer Flask
Click to view available options
Capacity (Metric):
100 mL
200 mL
300 mL
Unit Size:
Each
Description
- Graduated
- Transparent for easy viewing
- Non-stick PFA surface-easy to clean
- Can be used in a microwave
Specifications
Specifications
| Color | Translucent |
| Capacity (Metric) | 200 mL |
| Material | PFA |
| Height (Metric) | 123 mm |
| No. per Pack | 1 Ea. |