Learn More
Saint Gobain Life Sciences Chemware™ PTFE Flouropolymer Film
Product Code. 12347008
Change view
Click to view available options
Length (English):
25 ft.
Unit Size:
7.5m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
12347008 25 ft. 7.5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12347008 Supplier Saint Gobain Life Sciences Supplier No. D1069279
  • ONLINE EXCLUSIVE PRICE
    211.94€
    129.09€ / 7.5m
    Save 82.85€
    39% Off
Estimated Shipment: 14-01-2027
Chemware™ PTFE Flouropolymer Film ideal for lining or covering shelves, drawers, tables, trays, drain boards, and sinks.
  • Temperature and corrosion resistant
  • Non-stick surface
  • Non-porous and non-absorbent
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 25 ft.
Product Type Film
Width (English) 12 in.
Width (Metric) 305 mm
Length (Metric) 7.6 m
Material PTFE
Thickness 127 μm