Learn More
Esculape Laboratoire™ Chemistry First Aid Kit
Product Code. 11779315
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11779315 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11779315 Supplier Esculape Laboratoire™ Supplier No. 722 06 08
  • ONLINE EXCLUSIVE PRICE
    179.55€
    98.19€ / Pack of 1
    Save 81.36€
    45% Off
Estimated Shipment: 12-11-2026
Specifications
Depth (Metric) 10 cm
For Use With (Application) Environments Exposed to Chemical Hazards
Length (Metric) 27 cm
Product Type Chemistry First Aid Kit
Includes 1 pH neutral eye wash 200mL, 10 single dose saline, 1 survival blanket, one face shield mouth to mouth, 1 triangular scarf, 10 sterile compresses 30 x 30cm, 18 adhesive bandages assorted, 4 stretch bands 3m x 7cm, 2 compression bandages, 1 roll of tape 5m
Quantity 1 Ea.
Height (Metric) 29 cm