missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiScreen™ SST3 Somatostatin Receptor Membrane Preparation

Product Code. 17385309
Encompass
Change view
Click to view available options
Quantity:
1 Vial (200 units)
Unit Size:
1 ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17385309 1 Vial (200 units) 1 ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17385309 Supplier DiscoverX Supplier No. HTS171M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChemiScreen™ SST3 Somatostatin Receptor Membrane Preparation allows measurement of ligand binding to SSTR3 GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.

The ChemiScreen™ SST3 Somatostatin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of SSTR3 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.

Specifications

Assay Entry SST3
For Use With (Application) Membrane Preps
Quantity 1 Vial (200 units)
Target Entry SSTR3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.