missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiScreen™ NOP/ORL1 Opioid Receptor Stable Cell Line

Product Code. 17395039
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17395039 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17395039 Supplier DiscoverX Supplier No. HTS040C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChemiScreen™ NOP/ORL1 Opioid Receptor Stable Cell Line measures OPRL1 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.

The ChemiScreen™ NOP/ORL1 Opioid Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. orphanin FQ) based activation of OPRL1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses OPRL1 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of OPRL1, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.

Spécification

Assay Entry Calcium Flux
Cell Line Chem-1
For Use With (Application) Cell Lines
Primary Agonist Orphanin FQ
Quantity 2 Vials
Target Entry OPRL1
Accession Number AY268428
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.