missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiScreen™ NMU1 Neuromedin U Receptor Stable Cell Line

Product Code. 17315099
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17315099 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17315099 Supplier DiscoverX Supplier No. HTS062C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChemiScreen™ NMU1 Neuromedin U Receptor Stable Cell Line measures NMU1R (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.

The ChemiScreen™ NMU1 Neuromedin U Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. Neuromedin U) based activation of NMU1R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NMU1R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of NMU1R , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Calcium Flux
Cell Line Chem-1
For Use With (Application) Cell Lines
Primary Agonist Neuromedin U-25
Quantity 2 Vials
Target Entry NMU1R
Accession Number NM_006056
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.