missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX ChemiScreen™ MRGPRX2 Receptor Stable Cell Line
Description
The ChemiScreen™ MRGPRX2 Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cortistatin-14, PAMP) based activation of MRGPRX2 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses MRGPRX2 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of MRGPRX2, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Calcium Flux |
| Cell Line | Chem-1 |
| Format | 2 vials |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Proadrenomedullin fragment, 1-20 (PAMP) |
| Quantity | 2 Vials |
| Target Entry | MRGPRX2 |
| Accession Number | NM_054030.2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?