missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX ChemiScreen™ LPA3 Lysophospholipid Receptor Stable Cell Line

Product Code. 17385249
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17385249

Brand: DiscoverX HTS130C

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

The ChemiScreen™ LPA3 Lysophospholipid Receptor Stable Cell Line measures LPAR3 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.

The ChemiScreen™ LPA3 Lysophospholipid Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. LPA) based activation of LPAR3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses LPAR3 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of LPAR3 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Calcium Flux
Cell Line Chem-1
For Use With (Application) Cell Lines
Primary Agonist 1-Oleoyl-LPA
Quantity 2 Vials
Target Entry LPAR3
Accession Number NM_012152.2
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.