missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX ChemiScreen™ CXCR1 Chemokine Receptor Membrane Preparation
Description
The ChemiScreen™ CXCR1 Chemokine Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CXCR1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.
Specifications
Specifications
| Assay Entry | CXCR1 |
| For Use With (Application) | Membrane Preps |
| Quantity | 1 Vial |
| Target Entry | CXCR1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?