Learn More
DiscoverX ChemiScreen™ CB1 Cannabinoid Receptor Membrane Preparation
Product Code. 17384989
Encompass
Click to view available options
Quantity:
1 Vial
Unit Size:
1 ampoule
Product Code. 17384989
Brand: DiscoverX HTS019M
The ChemiScreen™ CB1 Cannabinoid Receptor Membrane Preparation allows measurement of ligand binding to CNR1 GPCR via radioligand binding assays, or GPCR activity via GTPγS assay. Membrane preps contain 1mL of 1mg/mL frozen membranes.
The ChemiScreen™ CB1 Cannabinoid Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CNR1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach or to screen for GPCR activity using GTPγS assays. Increased binding affinities or GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.
Specifications
Assay Entry CB1
For Use With (Application) Membrane Preps
Quantity 1 Vial
Target Entry CNR1
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.