Learn More
Thermo Scientific™ Cheminert Model C2 Microbore Injector
Product Code. p-3755832
Click to view available options
Volume (Metric) Sample:
10 μL
Unit Size:
Each
Product Code. 11523970
Brand: Thermo Scientific™ CZSL10PK
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Can be used as an injector or switching valve
  • 1/16 in. fittings
  • 0.010 in. ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
Specifications
Volume (Metric) Sample 10 μL