missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Chemical Test Kit
Click to view available options
For Use With (Application):
Free Chlorine LR/MR Determination
Free and Total Chlorine Determination
Iron (Fe2+ and Fe3+) Determination
Nitrite Determination
Phenols Determination
Phosphate Determination
Sulfite Determination
Total Chlorine Determination
Total Hardness Determination
Unit Size:
Each
Pack of 100
Pack of 50
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
| Analytes | Ascorbic acid |
| For Use With (Application) | Phosphate Determination |
| For Use With (Equipment) | Water Samples |
| Includes | 20mL plastic beaker, color comparison cube, and 50 packets phosphate reagent |
| No. of Tests | 50 |
| Quantity | 1 Kit |
| Range | 0 to 5mg/L |
| Test Method | Titration |
| Type | Phosphate Test kit |
Safety and Handling
Product Identifier
- Phosphate Reagent
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/first aider.
- P404-Store in a closed container.
- MIXTURE LIST-Contains : potassium disulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction