Learn More
Hanna Instruments™ Chemical Test Kit
Product Code. 11612310
Click to view available options
For Use With (Application):
Free Chlorine LR/MR Determination
Free and Total Chlorine Determination
Iron (Fe2+ and Fe3+) Determination
Nitrite Determination
Phenols Determination
Phosphate Determination
Sulfite Determination
Total Chlorine Determination
Total Hardness Determination
Unit Size:
Each
Pack of 100
Pack of 50
Product Code. 11612310
Brand: Hanna Instruments™ HI38020
Only null left
Add to basket
Add to basket
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Analytes DPD
For Use With (Application) Free and Total Chlorine Determination
For Use With (Equipment) Water sampples
Includes HI93701-0 free chlorine reagent (100 packets), HI93711-0 total chlorine reagent (100 packets), demineralizer bottle with filter cap for 12L, Checker disc, glass vials with caps (2), and 1 (3mL) plastic pipette
No. of Tests 100 ea. Free/Total (200 total)
Quantity 1 Kit
Range 0.00 to 0.70mg/L, 0.0 to 3.5mg/L, 0.0 to 10.0mg/L
Sufficient For Low, medium and high range determination
Test Method Titration
Type Chemical Test Kit
compliance-icons compliance-icons
Product Identifier
  • FREE & TOTAL CHLORINE LOW RANGE, M
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Purolite, Potassiumiodide, EDTA disodium salt, N,N-Diethyl-1,4-phenylenediammonium sulphate