missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX ChemiBrite™ GPBA Bile Acid Receptor Stable Cell Line
Description
The ChemiBrite GPBA Bile Acid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. lithocholic acid, other bile acids) based activation of GPBAR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPBAR1 alongside exogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of GPBAR1 , the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Calcium Flux |
| Cell Line | 293H |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Lithocholic Acid |
| Quantity | 2 Vials |
| Target Entry | GPBAR1 |
| Accession Number | NM_001077194.1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?