Learn More
Medimark International™ Chemgene HLD4L Laboratory Surface Disinfectant
Product Code. p-8000871
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
Product Code. 15136686
Brand: Medimark International™ XTM305
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Combines enhanced active ingredient molecules with micelle cleaning technology to provide swift penetration of the cell walls to ensure rapid cell death rather than merely suspending activity.
For multipurpose surface disinfection and cleaning. Chemgene™ Lab Disinfectant Spray destroys bacteria, fungi, mycobacteria, viruses and spores.
Chemgene HLD4L disinfectant concentrate.
  • Manufactured in accordance with Quality Management System ISO 9001
  • Biodegradable under OECD conditions
  • Denatures DNA/RNA
Specifications
Product Type Lab Disinfectant
Appearance Clear
Dilution 0.048611111111
For Use With (Application) For multipurpose Surface disinfection and cleaning
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • CHEMGENE HLD4L CONCENTRATE
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P315-Get immediate medical advice/attention.