missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics CHEMets Chlorine Free & Total Water Testing Kit

Product Code. 11767433
Click to view available options
Range:
0 to 1 ppm and 1 to 5 ppm
0.1 to 2 ppm
Test Method:
DDPD
DPD
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 11767433

Brand: CHEMetrics K2500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

REMOVE

Provides results in two minutes or less

Provide results in two minutes or less

  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life

Specifications

Analytes Free and Total Chlorine
Includes Plastic case, 30 ampoules of reagent, comparator(s), snap cup (holds water sample) and instructions
No. of Tests 30
Range 0.1 to 2 ppm
Test Method DPD
Type CheMets Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • CHEMETS CHLORINE 0-2 MG/L
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : acetone, potassium phosphate, N,N-diethyl-p-phenylenediamineoxalate, potassiumiodide, sodium hydro
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.