Learn More
CHEMetrics CHEMets Dissolved Oxygen, Visual Water Testing Kit
Product Code. 11731004
Change view
Click to view available options
Range:
0 to 1 ppm
0 to 100 ppb
0 to 40 ppb
1 to 12 ppm
Unit Size:
Pack of 30
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11731004 0 to 40 ppb Pack of 30
11711004 0 to 1 ppm Pack of 30
11721004 1 to 12 ppm Pack of 30
11781004 0 to 100 ppb Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11731004 Supplier CHEMetrics Supplier No. K7540
  • ONLINE EXCLUSIVE PRICE
    64.12€
    59.66€ / Pack of 30
    Save 4.46€
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life.
Specifications
Analytes Dissolved Oxygen (DO)
Includes Plastic case, Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions
No. of Tests 30
Range 0 to 40 ppb
Test Time 2 min.
Type CHEMets Visual Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • Dissolved Oxygen Test Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician/first aider.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.