Learn More
CHEMetrics CHEMets Dissolved Oxygen Test Kit Refill
Product Code. p-4443021
Click to view available options
Range:
0 to 1 ppm
0 to 40 ppb and 0 to 100 ppb
1 to 12 ppm
Test Method:
Indigo Carmine
Rhodazine D
For Use With (Equipment):
C-7501 comparators
C-7512 comparators
C-7540 comparators
Unit Size:
Pack of 30
Product Code. 11731044
Brand: CHEMetrics R7501
Only null left
Add to basket
Edge
Add to basket
Provides results in two minutes or less
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life
Specifications
Analytes Dissolved Oxygen (DO)
For Use With (Equipment) C-7501 comparators
Includes 30 self-filling CHEMets ampoules to refill kit K-7501
No. of Tests 30
Range 0 to 1 ppm
Test Method Rhodazine D
Test Time 2 min.
Type CHEMets Dissolved Oxygen Refill
compliance-icons compliance-icons
Product Identifier
  • Oxygen, dissolved
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.