Learn More
CHEMetrics CHEMets Silica Test Kit Refill
Product Code. 11721064
Change view
Click to view available options
Range:
0 to 1 ppm and 1 to 10 ppm
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11721064 0 to 1 ppm and 1 to 10 ppm Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11721064 Supplier CHEMetrics Supplier No. R9010
 Restricted Item
Estimated Shipment: 02-10-2026
Provides results in two minutes or less
This product contains 30 self-filling CHEMets ampules to refill kit K-9010 for the analysis of silica. R-9010 is compatible with C-9001 and C-9010 comparators.
Specifications
Analytes Silica
For Use With (Equipment) C-9001 and C-9010 comparators
Includes 30 self-filling CHEMets ampoules to refill kit K-9010
Range 0 to 1 ppm and 1 to 10 ppm
Test Method Heteropoly Blue
Type Chemets™ Refill
compliance-icons
Product Identifier
  • refill CHEMets SiO2 0-10PPM
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P271-Use only outdoors or in a well-ventilated area.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: diethylene glycol, sodium bisulphate, 1-amino-2-napthol-4-sulphonic acid, sodiumsulphite