Learn More
Masterflex™ Chem-Durance Bio L/S™ Two-Stop Pump Tubing
Product Code. 16438743
Click to view available options
Tubing Size:
13
16
25
Unit Size:
Pack of 8
Product Code. 16438743
Brand: Masterflex™ 9611716
Only null left
Add to basket
Add to basket
Excellent chemical resistance and long pumping life
  • Meets USP Class VI requirements
  • Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers - extending the life of the tubing
  • Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids
Specifications
Diameter (Metric) Inner 3.1 mm
Material Chem-Durance Bio
Overall Length (English) 1.25 ft.
Overall Length (Metric) 0.38 m
For Use With (Application) Laboratory, Pharmaceutical Processing, and Biotech
Min. Operating Temperature (Metric) 0°C
Packaging Quantity 8
Tubing Size 16
Flow Rate 460 mL/min
Hose Barb 1/8 in.
Maximum Pressure 40 psi, 2.7 bar
Vacuum Level 660 mm Hg
Max. Operating Temperature (Metric) 40°C
Product Type 2-stop Tubing